Tlačiť
Swabs for Protein Detection
Dodávateľ: HyServe
SCIL1402954EA 1836.24 EUR
SCIL1402954 710-2025 710-2026
Swabs for Protein Detection
Tampóny odběrové Tampóny pro odběr vzorků
Sterile swabs in tube for sampling. The swab is returned to the tube and the sample is extracted in 1 ml of buffer solution.
  • Storage at room temperature for up to two years
  • Can be dispensed on to a compact dry or a petri dish
Swabs for sampling in difficult areas and checking dry areas.
Pozor: Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904