Tlačiť
Disposable needles for dental anaesthesia, Sterican®
Dodávateľ: B. Braun
720-2562EA 17.74 EUR
720-2562 BRAU9186158 720-2565 720-2566 720-2563 720-2564
Disposable needles for dental anaesthesia, Sterican®
Jehly ke stříkačkám
Needle tube is made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating.
  • Thin walled needle with three facet bevel for minimal pain upon puncture
  • Long bevel for easy venipuncture of normal veins
  • Transparent, colour-coded, Luer lock plastic hub made from PP
Certifikácie: ISO 7864
Balenie: In a dispenser box.