Tlačiť
Paraffin section mounting bath
Dodávateľ: Electrothermal
114-8736EA 1496.13 EUR
ELETMH8517
Paraffin section mounting bath
This bath is designed to assist with the handling of paraffin wax samples in histology and pathology laboratories. It is essentially a hot distilled water floating-out bath that allows for the meticulous manipulation and location of paraffin wax sections onto glass slides. A white neon light indicates 'Power On' and an amber neon light indicates 'Heater On'. The required temperature is selected using the heater dial. Accurate water temperature is then maintained through an energy regulator which supplies power to a silicone rubber mat in controlled, timed pulses.
  • Aluminium base with PTFE coated interior
  • Virtually maintenance-free and extremely easy to clean
  • Excess or damaged sections can simply be removed by skimming a filter paper over the water surface
  • 70 °C nominal temperature, for 2 litres of water, is attained within 120 minutes
  • Over-heating is prevented by a thermal fuse within the heater mat which automatically cuts out if too high a temperature is reached
Informace o dodávce: Supplied with an IEC moulded cord, lead set and the IEC socket houses protection fuses for both the live and neutral power supply lines. The lid needs to be ordered separately.