Tlačiť
Hľadali ste: Séra
104  results were found
SearchResultCount:"104"
Sort Results
Zobraziť zoznam Easy View (Nové)
Ohodnotiť tieto výsledky vyhľadávania
Dodávateľ: VWR Seradigm
Opis: Avantor Seradigm Select Grade FBS provides superior performance and consistency in cost-sensitive applications.
Dodávateľ: VWR Seradigm
Opis: Australia Origin FBS is widely considered the gold standard for safety and regulatory compliance by cell culturists around the world. Avantor Seradigm...
Dodávateľ: VWR Seradigm
Opis: Ultimate grade FBS offers the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology. Premium gra...
Dodávateľ: VWR Seradigm
Opis: Ultra-low IgG foetal bovine serum (FBS) is purified of immunoglobulin (IgG) using a chromatographic process that delivers significantly reduced levels...
Dodávateľ: BIOWEST
Opis: High quality, sterile filtered foetal bovine serum (FBS) derived exclusively from EU countries. Each manufactured batch is rigorously controlled, from...
Katalógové číslo: (IMMRIR2420)
Dodávateľ: IMMUNOREAGENTS
Opis: Normal bovine serum is used as a blocking agent or as a control for immunoassays. All serum is obtained from healthy, disease free animals. Serum is l...
Merná jednotka: 1 * 10 mL
Dodávateľ: BIOWEST
Opis: Serum, Zdroj séra: Animal, Fetální hovezí sérum, Původ FBS: Australia, USDA approved, Frozen, Sterile filtered
Dodávateľ: BIOWEST
Opis: Rat serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens remo...
Dodávateľ: BIOWEST
Opis: Feline serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens r...
Dodávateľ: HyClone products (Cytiva)
Opis: HyClone™ donor horse serum, designed to supplement cell growth, is collected from selected donor herds that receive regular veterinary inspection and ...
Katalógové číslo: (HYCLSH30212.03)
Dodávateľ: HyClone products (Cytiva)
Opis: HyClone™ Alpha Calf Fraction provides hybridoma users with a cost-effective medium supplement that promotes growth and antibody production.
Merná jednotka: 1 * 500 mL
Dodávateľ: HyClone products (Cytiva)
Opis: Highest quality FBS, HyClone US-origin defined FBS is 40 nm-filtered, virus-tested to 9 CFR standard, and supplied with an extensive biochemical profi...
Dodávateľ: HyClone products (Cytiva)
Opis: All New Zealand calf serum products are carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases. We follo...
Dodávateľ: HyClone products (Cytiva)
Opis: This premium HyCloneTM Fetal Bovine Australian-origin Serum is well-characterized, triple 100 nm filtered, virus-tested to 9 CFR standard, and traceab...
Dodávateľ: BIOWEST
Opis: Guinea pig serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinoge...
Dodávateľ: BIOWEST
Opis: Sterile filtered foetal bovine serum (FBS) derived from clotted whole blood aseptically collected from foetus via cardiac puncture. Each manufactured ...
Zavolať a opýtať sa na ceny
Zásoby tejto položky sú obmedzené, no môžu sa nachádzať v inom sklade blízko pri vás. Uistite sa prosím, že ste prihlásený/á na tejto lokalite, aby sa mohli zobraziť dostupné zásoby. Ak sa stále zobrazuje zavolajte a potrebujete pomoc, zavolajte nám prosím na číslo +43 1 97002 - 0.
Zásoby tejto položky sú obmedzené, no môžu sa nachádzať v inom sklade blízko pri vás. Uistite sa prosím, že ste prihlásený/á na tejto lokalite, aby sa mohli zobraziť dostupné zásoby. Ak sa stále zobrazuje zavolajte a potrebujete pomoc, zavolajte nám prosím na číslo +43 1 97002 - 0.
Daná chemikália je regulovaná podľa platnej legislatívy a bude vyžadované vyplnenie formulára. Jeho včasným vyplnením urýchlite dodanie produktu.
-Na zakúpenie tejto položky môže byť potrebná dodatočná dokumentácia. Ak to bude potrebné, skontaktuje sa s vami zástupca spoločnosti VWR.
Tento produkt bol zablokovaný vašou organizáciou. Kontaktujte prosím vaše oddelenie nákupu pre získanie viac informácií.
Pôvodný produkt už nie je dostupný. Je dostupná zobrazená náhrada.
Tento produkt už nie je dostupný. Môžu byť k dispozícii alternatívy, ktoré sa dajú vyhľadať pomocou katalógového čísla VWR uvedeného vyššie. Ak potrebujete ďalšiu pomoc, zavolajte prosím na zákaznícky servis spoločnosti VWR na čísle +43 1 97002 - 0.
1 - 16 of 104
no targeter for Bottom