Tlačiť
Corning® Human Extracellular Matrix
Dodávateľ: Corning
734-0097EA 287.78 EUR
BDAA354237
Corning® Human Extracellular Matrix
Human extracellular matrix (ECM) is a chromatographically partially purified matrix extract derived from human placenta. It is comprised of collagen IV, laminin and heparan sulphate proteogycan. Human ECM promotes attachment, spreading, mitosis, and differentiation of anchorage-dependent epithelial cells, particularly of human origin.