Tlačiť
dNTP mix
Dodávateľ: Thermo Fisher Scientific
733-0548EA 91.48 EUR
733-0548
dNTP mix
Činidla pro nukleové kyseliny Nukleotidy
A combined mixture of  dATP, dCTP, dGTP and dTTP containing 5 mM of each dNTP. Applications include PCR, RT-PCR, DNA labelling, and site-directed mutagenesis.