Tlačiť
Barcode reader, SingleScan™, WHEATON®
Dodávateľ: Wheaton
SingleScan™
479-1146EA 4342.02 EUR
479-1146
Barcode reader, SingleScan™, WHEATON®
Systémy na čárové kódy
This barcode reader for 2D data matrix and 1D linear barcodes enables users to identify and track their samples with ease.
  • USB interface allows easy connection to computer
  • Plug and play design enables easy set-up with no software installation required
  • Inputs decoded barcode ID into any software application where cursor is placed
The SingleScan is capable of reading a variety of barcoded vials or ampoules from a wide range of manufacturers and label printers. Barcodes should adhere to the ISO16022 Data Matrix Two Dimensional Barcode format and ISO22742 Linear Barcode and two dimensional symbols for product packaging.