Tlačiť
Pipette tips, Matrix®
Dodávateľ: Thermo Fisher Scientific
Matrix®
613-1836EA 164.6 EUR
613-1836 613-1858
Pipette tips, Matrix®
Špičky pipetovací
Transparent PP, sterile or non sterile.
  • Sealing rings on tips ensure confident, leak-free seal on most pipettes
  • Tip manufactured under tight QC specifications ensures low liquid retention without special treatments or additives
  • Extended tips allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the pipette barrel against the side of the tube
Balenie: Available bulk, in Flo-Thru racks or in Ecotips refills. Flo-Thru racks individually support tips to prevent them from wobbling when trying to seat them on the pipette. Ecotips refill system is 100% recyclable and offers fast rack reloading in a space saving design.