Tlačiť
Bottle carrier for B.O.D. bottles, SP Bel-Art
Dodávateľ: Bel-Art Products, a part of SP
SP Bel-Art
BELAF169680000EA 149.08 EUR
BELAF169680000
Bottle carrier for B.O.D. bottles, SP Bel-Art
Přepravky Přepravky na láhve/nádoby
Bottle carrier made of polypropylene (PP), for eight standard 300 ml Biological Oxygen Demand bottles.
  • Tall handle for easy carrying
  • Openings are 72 mm diameter
  • Steam atuclavable at 121 °C