Chcete sa opäť prihlásiť?
Inspissator for the production of Tuberculosis culture medium
Dodávateľ: Grant
This convenient and effective system was developed in conjunction with the Royal Postgraduate Medical School of London University to produce large batches of uniform tuberculosis culture medium four to six times per day. Vessels containing culture medium are incubated on a shallow tray which is in contact with water held at a constant temperature of 85 ºC within a tank, ensuring that the temperature of the vessels is constant. Inspissation takes 50 minutes at 85 ºC. It is robust and durable as the tray, tank and outer case are made of polished stainless steel. Water temperature under the tray is controlled by a digital immersion thermostat for accuracy and reproducibility of set temperature. All moving parts are incorporated in the control unit which is easily removed for cleaning. An insect resistant blanket and quilt are placed over the containers to provide thermal insulation and exclude draughts.
  • Robust durable design, with digital temperature control and clear 4 digit display which is easy to read from a distance
  • Standard temperature: 85ºC; temperature range ambient +5 to 100 ºC
  • Capacity for up to 156 test tubes (16 mm diameter x 150 mm long) or 162 universal containers
  • Constant level device maintains required liquid level for optimal operation and a fixed over temperature cutout protects users, valuable samples and the workplace
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
TBT-T100IN, for tuberculosis culture medium, holds 156 test tubes or 162 universal containers 1 GRANTBT-T100IN
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Heater power (W) 1300
Uniformity (°C) ±0,7
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo