Tlačiť

Agaróza, II™, for biotechnology

0815-25GEA 340.28 EUR
0815-25G 0815-250G 0815-100G
Agaróza, II™, for biotechnology
Agaróza

A low melting agarose that melts between 62 to 68 °C and remains liquid for several hours at 37 °C. Ideal for purification of macromolecules from agarose slices and for in-gel enzymatic manipulations.

Teplota tavenia: 60-90 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00081294
Číslo CAS: 9012-36-6
EINECS: 232-731-8
https://sk.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Výsledky špecifikačného testu

DNase NONE
EEO (-Mr) ≤0.12
Gel strength (1.0%) ≥250 g/cm²
Gelling Range (1.0%) 24 - 29 °C
Melting range 62 - 68 °C
Moisture <10%
Protease NONE
RNase NONE
Sulphate ≤0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR