Tlačiť
Reflectance colorimeters, RT 300, RT 400, RT 500, Lovibond®
Dodávateľ: Tintometer
633-0250EA 12688.88 EUR
633-0250 633-0248 633-0247 633-0249
Reflectance colorimeters, RT 300, RT 400, RT 500, Lovibond®
Kolorimetry Kolorimetry reflexní
The RT300, RT400 and RT500 are versatile, portable spectrophotometers in a choice of diffuse/8° sphere or 0/45° optical geometry that are designed to give fast, precise and accurate colour measurement information on a range of products. They provide absolute and difference measurements in various colorimetric systems.
The RT300 and RT500 can be combined with the OnColor™ software package allowing measurement data to be downloaded, analysed in detail and stored indefinitely on a PC.
  • Lightweight, compact portable instruments
  • Opacity and colour strength measurement
  • Data storage for 1024 standards with tolerances and 2000 samples
  • Flip-back target shoe for flexible use
  • Rechargeable battery for remote use
Informace o dodávce: Supplied with calibration standards, AC adapter, carrying case and operation manual. Other models are available, please contact your local Avantor Sales Office for further details.