Tlačiť
Test tubes, round bottom, soda glass, Sterilin®
Dodávateľ: Thermo Scientific
Sterilin®
SLIN49684EA 0 EUR
SLIN49684
Test tubes, round bottom, soda glass, Sterilin®
Zkumavky Zkumavky na vzorky
Soda-lime glass.
  • Provides greater chemical resistance than plastic
  • Test tube profile with rim
  • Supplied without caps
  • Not available in all geographic regions
Rely on these round base glass tubes for excellent chemical resistance for samples to 2 or 4 ml. Thermo Scientific™ Sterilin™ round base soda glass tube profile enables streamlined handling and excellent mixing capability. Not available in America.