Tlačiť
Tablet sampler
Dodávateľ: Sampling Systems
SAMP4010A-01EA 3805.19 EUR
SAMP4010A-01
Tablet sampler
Designed to take large samples of tablets, pellets, capsules and granules from a container as the product receiving container is filled.
Pozor: Place the sampler in the container before it is filled.