Tlačiť
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Dodávateľ: Thermo Fisher Scientific
ACRO333915000EA 256.7 EUR
ACRO333915000
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Proteiny
  • Infrared spectrum: Conforms
  • Specific optical rotation: –14 to –18° (20 °C, 589 nm) (C=1, 1 N HCl)
  • EINECS: 261-529-2
  • GHS symbol and signal word: GHS07 and warning