Tlačiť
ICNA02101927
Dodávateľ: MP Biomedicals
ICNA0210192791EA 1004.45 EUR
ICNA0210192791
ICNA02101927
HEPPS is the propane analog of HEPES.
HEPPS is used as a separator in ultrathin isoelectric focusing gels; enhances the resolution of phosphoglucomutase. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration (produced by cellular respiration).
  • Presentation: White Powder
  • Soluble in water (1 M - clear, colorless solution).