Tlačiť
Flip-top vials specific for aseptic sampling
Dodávateľ: Capitol Europe
216-4909EA 121.06 EUR
216-4909 216-4910
Flip-top vials specific for aseptic sampling
Vialky
PP, translucent, aseptic, single-use sample vials for field collection of farm milk and related dairy products, veterinary specimen collection, infection control of mastitis and environmental sampling. Very efficient during storage and transport. Suitable for food contact.
  • Manufactured in a cleanroom
  • Proprietary 'in-mould' closing process
  • Can be supplied on request with bronopol and/or barcode label
Certifikácie: Alimentary: Complies with CE norm no. 1935/2004
Balenie: A label on each box includes batch number and production date for traceability.
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904