Tlačiť
Flip-top vials for dairy herd improvement testing
Dodávateľ: Capitol Europe
CAPI03028071SHBPCNEA 289.04 EUR
CAPI03028071SHBPCN
Flip-top vials for dairy herd improvement testing
Vialky
Translucent PP. Aseptic, single-use sample vials for field collection of cow milk for mastitis evaluation and DHIA quality control. Suitable for food contact.
  • Manufactured in a cleanroom
  • Proprietary 'in mould closing' process
  • Can be supplied with bronopol and barcode label
Certifikácie: Complies with CE norm No.1935/2004
Balenie: A label on each box includes batch number and production date for traceability.
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904