Tlačiť

Iminodibenzyl

MOLEM24565519EA 0 EUR
MOLEM24565519
Iminodibenzyl
Iminodibenzyl
Vzorec: C₁₄H₁₃N
Mw: 195,27 g/mol
Číslo MDL: MFCD00005070
Číslo CAS: 494-19-9
EINECS: 207-787-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR