Tlačiť
Organic size exclusion chromatography columns, GPC KF-800 series preparative type
Dodávateľ: Resonac
SHOWF6108025EA 22265.38 EUR
SHOWF6108025 SHOWF6108030 SHOWF6108040 SHOWF6108010 SHOWF6108020 SHOWF6700408
Organic size exclusion chromatography columns, GPC KF-800 series preparative type
Kolony chromatografické
These are styrene divinylbenzene copolymer-based organic solvent SEC (GPC) columns. Suitable for a wide range of applications from low to high molecular weight compounds.