Tlačiť
Hydrophobic interaction chromatography column, HIC PH-814
Dodávateľ: Showa Denko
554-2852EA 1204.7 EUR
554-2852
Hydrophobic interaction chromatography column, HIC PH-814
Kolony chromatografické
  • Separates proteins without denaturation
  • Applicable to samples obtained after ammonium sulphate fraction treatment
  • Polyhydroxymethacrylate with Phenyl (functional group)