Tlačiť
Back supports, ErgoPerfect®
Dodávateľ: ErgoPerfect
ERGO4100012 ERGO4100015 ERGO4100013 ERGO4100014
Back supports, ErgoPerfect®
These supports are ideal for use during work and other physical activities. The unique properties of the neoprene blend provide compression, support and heat retention, which improves circulation, helps keep muscles and joints flexible and promotes healing.
  • Provides back support, compression and heat retention
  • Full coverage of back
  • Extra back support built-in