Tlačiť
Instrument trays with lid
Dodávateľ: Hecht
HECH42911004EA 71.22 EUR
HECH42911004
Instrument trays with lid
Glass, cover with foam plate and weight (glass plate).
  • Resistant to hot-air sterilization up to +180 °C
  • Excellent chemical resistance
  • Easy to clean