Tlačiť
Vacuum controller, VC 2 IKAVAC®
Dodávateľ: IKA
IKAVAC®
IKAA2300000
Vacuum controller, VC 2 IKAVAC®
Čerpadla Kontroléry vakua
Vacuum controller used to create a controlled partial vacuum in laboratory applications. For typical tasks such as evacuation of desiccators, vacuum apparatus, etc.
  • 99% Solvent recovery
  • Integrated air release valve
  • Space-saving stand-supported instrument
  • Automatic setpoint correction
  • Clearly organised membrane keyboard