Tlačiť

Disodium maleate

Dodávateľ: MP Biomedicals

ICNA0521804650EA 113.72 EUR
ICNA0521804650
Disodium maleate
Disodium maleate
Vzorec: C₄H₂Na₂O₄
Číslo MDL: MFCD00013059
Číslo CAS: 371-47-1
EINECS: 206-738-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR