Tlačiť
Gel documentation system, Syngene InGenius³
Dodávateľ: Synoptics
733-2313EA 4561.41 EUR
733-2313
Gel documentation system, Syngene InGenius³
Systémy fotodokumentační
Syngene InGenius³ is an ideal choice for low budget gel documentation and analysis for fluorescence applications. It features a compact darkroom which has a sliding front door and an internal LED white light.
  • High resolution CCD digital camera with manual zoom 6,5 to 39 (f/1,4)
  • Captured images can be stored and enhanced using the software supplied
  • Images can be saved onto PC via Ethernet or USB interface in JPEG, TIFF, or BMP formats
  • Darkroom with large access door and safety interlocks, built-in white light and filter drawer for interchangeable filters
  • Maximum sample size 200×200 mm
InGenius³ has a 3 megapixel camera and can produce images with 12 or 16 bit image depth.
Informace o dodávce: The system includes a 3 megapixel CCD camera, manual zoom lens and manual filter drawer (includes a UV filter), and epi-LED white lights. GeneSys image capture and manipulation software and 2× GeneTools analysis software are also supplied. An external PC, optimised for Windows 7 Pro is required (not supplied). A UV transilluminator is also required (not supplied). Please contact your local VWR office for full details on the options and accessories available for this system.