Tlačiť

ICNA02150162

Dodávateľ: MP Biomedicals
ICNA0215016250EA 416.84 EUR
ICNA0215016250 ICNA0215016280
ICNA02150162
Hydroxylapatite binds double-stranded but not single-stranded nucleic acids in lower concentrations of phosphate buffer. Double-stranded nucleic acids are eluted with a more concentrated phosphate buffer.
Hydroxylapatite is used for purification of proteins, separation of single-stranded RNA from double-stranded RNA & purification of DNA.
Store at Room Temperature (15-30 °C)

  • Grade: Fast Flow
  • Presentation: White Powder
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR