Tlačiť
Milk testing pipettes
Dodávateľ: Greiner bio-one
612-1031EA 0 EUR
GREI600107_1000
Milk testing pipettes
Disposable demeter pipettes made of transparent PS for bacteriological examinations in the dairy products industry.
  • With graduations (scale 0,5 /1,0/1,1 ml)
  • With cotton plug
  • Free of detectable DNase/RNase, human DNA and pyrogens