Tlačiť
Detergent, stainless steel cleaner
Dodávateľ: Micronova
115-1819EA 178.42 EUR
115-1819
Detergent, stainless steel cleaner
Prostředky čistící Detergenty a povrchové čističe
Gel stainless steel cleaner uses advanced no-drip gel technology to eliminate streaks and drips when applied to stainless, metal and other hard surfaces.
  • 'Sprayed' gel clings on contact and glides over most surfaces with minimum effort
  • Gel can be applied to seams and crevices to pre-treat before wiping, when dealing with rust, oxidation and residue build up
  • Gel is not atomised on spraying into ambient air and so will not impact indoor air quality, no VOCs
  • Contains no phosphorus