Tlačiť
Goggles, Atom
Dodávateľ: Bollé Safety
Atom
111-0969EA 20.87 EUR
111-0969 111-0966 111-0968
Goggles, Atom
Brýle bezpečnostní
Compact, lightweight, ultra-comfortable safety goggles. Thanks to its TPR flexible frame, Atom provides high protection. Fits over prescription spectacles and with visor can be used as faceshield.
  • TPR (Thermo Plastic Rubber) frame
  • Adjustable, ball-pivoting strap
  • Anti-scratch, anti-fog coatings
  • With or without foam (depending on version)
  • Liquid overflow chute
  • Fits with respiratory half mask
Weight: 74 g
Certifikácie: EN 166, EN 170