Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Reaction vessels, cylindrical

Dodávateľ: LENZ LABORGLAS GMBH & CO.KG
DURAN®, borosilicate glass, clear, with round bottom.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With easily readable graduation and laboratory flat flange (LF) with groove.




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko

With flat bottom
Kapacita S Vonkajší Ø. Výška Pk Číslo v katalógu VWR Množstvo
100 ml - 65 mm 75 mm 1 LENZ06000137
250 ml - 65 mm 130 mm 1 LENZ06000149
250 ml - 105 mm 130 mm 1 LENZ06000249
500 ml - 65 mm 230 mm 1 LENZ06000158
500 ml - 105 mm 150 mm 1 LENZ06000258
1000 ml laboratory flange LF 100 105 mm 200 mm 1 LENZ06000270
2000 ml laboratory flange LF 100 105 mm 330 mm 1 LENZ06000273
2000 ml - 160 mm 200 mm 1 LENZ06000473
3000 ml - 160 mm 250 mm 1 LENZ06000476
4000 ml laboratory flange LF 100 160 mm 350 mm 1 LENZ06000279
4000 ml - 160 mm 325 mm 1 LENZ06000479
5000 ml - 160 mm 350 mm 1 LENZ06000482
6000 ml - 160 mm 382 mm 1 LENZ06000485
10000 ml - 250 mm 410 mm 1 LENZ06000488
10000 ml - 215 mm 430 mm 1 LENZ06000588
20000 ml - 250 mm 630 mm 1 LENZ06000494
20000 ml - 315 mm 440 mm 1 LENZ06000594
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping