Chcete sa opäť prihlásiť?
Reaction vessels, cylindrical
Dodávateľ: LENZ LABORGLAS GMBH & CO.KG
DURAN®, borosilicate glass, clear, with round bottom.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With easily readable graduation and laboratory flat flange (LF) with groove.
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
With flat bottom
Kapacita S Vonkajší Ø. Výška Pk Číslo v katalógu VWR Množstvo
100 - 65 75 1 LENZ06000137
250 - 65 130 1 LENZ06000149
250 - 105 130 1 LENZ06000249
500 - 65 230 1 LENZ06000158
500 - 105 150 1 LENZ06000258
1000 laboratory flange LF 100 105 200 1 LENZ06000270
2000 - 160 200 1 LENZ06000473
3000 - 160 250 1 LENZ06000476
4000 laboratory flange LF 100 160 350 1 LENZ06000279
4000 - 160 325 1 LENZ06000479
5000 - 160 350 1 LENZ06000482
6000 - 160 382 1 LENZ06000485
10000 - 250 410 1 LENZ06000488
10000 - 215 430 1 LENZ06000588
20000 - 250 630 1 LENZ06000494
20000 - 315 440 1 LENZ06000594
2000 laboratory flange LF 100 105 330 1 LENZ06000273
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo