Chcete sa opäť prihlásiť?
Centrifuge, refrigerated, floor-standing, Sorvall™ RC-6 Plus superspeed
Dodávateľ: Thermo Scientific
Sorvall®
The Sorvall™ RC-6 Plus superspeed centrifuge combines superior functionality, unparalleled safety and traditional Sorvall™ reliability in a compact, user-friendly design.
  • Non bolt-down design allowing the centrifuge to be placed within the lab without being anchored it to the floor
  • Intuitive controls and dual display
  • 30-program memory allowing common protocols to be stored and retrieved easily, and offering flexibility in a multi-user environment
  • GMP/GLP-compatible data management (Traceability and quality control for Biotech/Pharmaceutical processing needs)
Offering 4-liter capacity and speeds of up to 22 000 min⁻¹ (55 200×g), the Sorvall™ RC-6 Plus is a high-productivity solution that delivers maximum throughput and minimum maintenance.
Ordering information: Other rotors available on request.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Rotors
Opis Pk Číslo v katalógu VWR Množstvo
Fixed-angle rotor SLA-3000, aluminium, 6×500 ml, 11000 min-1, 20450×g 1 521-1609
Fixed-angle rotor SE-12, aluminium, 12×14 ml, 26000 min⁻¹, 7 450×g 1 521-1613
Fixed-angle rotor SS-34, aluminium, 8×50 ml, 20500 min⁻¹, 50230×g 1 521-1615
Fiberlite carbon fiber rotor F21S-8×50y, 8×50 ml, 20000 min-1, 47488×g 1 521-1620
Special purpose rotor SV-80, aluminium, 16×5 ml, 19000 min-1, 40,970 ×g 1 SORV09200
Swinging bucket rotor SH-3000, 4×750 ml, 4700 min-1, 4,575 ×g (buckets), 3,706 ×g (microplate carriers) 1 SORV11796
Swinging bucket rotor HB-6, 6×50 ml, 13000 min-1, 11,855 ×g 1 SORV11860
Fixed angle rotor, F-20/micro, 32×1,5 ml, 20000 min-1, 51,427 ×g 1 SORV27038
Fixed angle rotor SA-600, 12×50 ml, 17000 min-1, 41,837 ×g 1 SORV28500
Fixed-angle rotor, F13-14x50cy, 14×50 ml, 13000 min-1, 28960×g 1 SORV46922
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Model
Teplota (°C)
Max. capacity
Max. rýchlosť (min-1)
Max. RCF (×g)
Acceleration/braking time
Motor
Ovládanie
Programy
Časovač
ŠxH×V (mm)
Hmotnosť (kg)
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo