Chcete sa opäť prihlásiť?
Stepsaver™ 70 v6 8 x (2 x 2.0 ml), titanium, 70,000 rpm/467,400 x g (with complete accessories as listed in rotor catalogue) 1 * 1 items
Dodávateľ: SORVALL
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Popis Číslo v katalógu VWR Jednotka Počet
Stepsaver™ 70 v6 8 x (2 x 2.0 ml), titanium, 70,000 rpm/467,400 x g (with complete accessories as listed in rotor catalogue) 1 * 1 items SORV79274 1 * 1 items
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo